Water purification system, AFS® 10E/15E
AFS® 10E water purification system (EU plug, manual in Danish)
MILLZAFS10E0DK
Economical water system with Elix® technology, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 L/min. For clinical analysers with pure water needs up to 300 L daily.
- Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality
- Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
- No resin cylinders to replace: Lower running costs
- Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
- Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
Patented Elix® technology provides constant quality water without the need for softeners or conditioning cartridges, which means reduced maintenance― and less analyser downtime.
- Description:AFS® 10E water purification system (EU plug, manual in Danish)
- Seq:02
- Cat. No.:MILLZAFS10E0DK
- Production flow rate:10 L/h @ 25 °C
- Feed water:Portable tap water at a maximum pressure of 6 bar
- Model:AFE® 10E
- Bacteria:< 1 CFU/ml
- W×D×H:268×426×585 mm
- Resistivity at +25 °C:>15 MΏ.cm
- TOC content:< 30 ppb
- Weight:28 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



