Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Powder funnels, PP

Powder funnels, PP

100 mm, 26 mm
Catalog # SAMP8023P-100
Supplier:   Sampling Systems
Powder funnels, PP
Catalog # SAMP8023P-100
Supplier:  Sampling Systems
Powder funnels, PP
Powder funnels, PP

100 mm, 26 mm

Price
3,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.

  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs
Specifications
  • Funnel O-Ø:
    100 mm
  • Seq:
    01
  • Stem O-Ø:
    26 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.