Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, loc347273( np_001018126.1, 1 a.a. - 364 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00347273-P01
Supplier:  Abnova
Recombinant protein, loc347273( np_001018126.1, 1 a.a. - 364 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00347273-P01
Supplier:  Abnova