Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, gin1( aah15325.1, 1 a.a. - 222 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00054826-P01
Supplier:  Abnova
Recombinant protein, gin1( aah15325.1, 1 a.a. - 222 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00054826-P01
Supplier:  Abnova