Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, c10orf54( aah20568.1, 1 a.a. - 311 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00064115-P01
Supplier:  Abnova
Recombinant protein, c10orf54( aah20568.1, 1 a.a. - 311 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00064115-P01
Supplier:  Abnova