Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, snip( aah33233.1, 1 a.a. - 257 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00080725-P01
Supplier:  Abnova
Recombinant protein, snip( aah33233.1, 1 a.a. - 257 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00080725-P01
Supplier:  Abnova