Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, anks1( np_056060, 1037 a.a. - 1132 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00023294-Q01
Supplier:  Abnova
Recombinant protein, anks1( np_056060, 1037 a.a. - 1132 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00023294-Q01
Supplier:  Abnova