0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
CCT5 RECOMBINANT PROTEIN (P01) 1 * 1 EA
ABNOH00022948-P01
:
Abnova
CCT5 RECOMBINANT PROTEIN (P01) 1 * 1 EA
ABNOH00022948-P01
:
Abnova
Literature reference - PubMed ID 23620386
sds_ABNOH00022948-P01.pdf
TDS_ABNOH00022948-P01.pdf
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR®, Nitrilhandschoenen
: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH-teststrips, MQuant®, Supelco®
: Merck
VWR®, gezichtstissues
: VWR Collection