Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, pcbp3( aah12061.1, 1 a.a. - 313 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00054039-P01
Supplier:  Abnova
Recombinant protein, pcbp3( aah12061.1, 1 a.a. - 313 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00054039-P01
Supplier:  Abnova