Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, fbxo40( np_057382.1, 1 a.a. - 709 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051725-P01
Supplier:  Abnova
undefined
Recombinant protein, fbxo40( np_057382.1, 1 a.a. - 709 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051725-P01
Supplier:  Abnova