Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, ttrap( aah17553, 1 a.a. - 363 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051567-P01
Supplier:  Abnova
undefined
Recombinant protein, ttrap( aah17553, 1 a.a. - 363 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051567-P01
Supplier:  Abnova