Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, kif3a( aah45542.1, 1 a.a. - 702 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00011127-P01
Supplier:  Abnova
Recombinant protein, kif3a( aah45542.1, 1 a.a. - 702 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00011127-P01
Supplier:  Abnova