Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, tjp2( np_004808, 121 a.a. - 218 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009414-Q01
Supplier:  Abnova
Recombinant protein, tjp2( np_004808, 121 a.a. - 218 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009414-Q01
Supplier:  Abnova