Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, adam15( aah14566, 301 a.a. - 410 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008751-Q01
Supplier:  Abnova
Recombinant protein, adam15( aah14566, 301 a.a. - 410 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008751-Q01
Supplier:  Abnova