Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, eif3s2( np_003748.1, 226 a.a. - 325 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008668-Q01
Supplier:  Abnova
Recombinant protein, eif3s2( np_003748.1, 226 a.a. - 325 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008668-Q01
Supplier:  Abnova