Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, arfgef1( np_006412.2, 311 a.a. - 411 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010565-Q01
Supplier:  Abnova
Recombinant protein, arfgef1( np_006412.2, 311 a.a. - 411 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010565-Q01
Supplier:  Abnova