Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, sox4( np_003098, 45 a.a. - 136 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006659-Q01
Supplier:  Abnova
Recombinant protein, sox4( np_003098, 45 a.a. - 136 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006659-Q01
Supplier:  Abnova