Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, smarce1( np_003070, 75 a.a. - 142 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006605-Q01
Leverancier:  Abnova
undefined
Recombinant protein, smarce1( np_003070, 75 a.a. - 142 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006605-Q01
Leverancier:  Abnova
Gerelateerde informatie
Frequently Bought Together