Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, myh9 ( np_002464.1, 1747 a.a. - 1852 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004627-Q02
Supplier:  Abnova
Recombinant protein, myh9 ( np_002464.1, 1747 a.a. - 1852 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004627-Q02
Supplier:  Abnova