Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, tspan8( np_004607, 110 a.a. - 205 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007103-Q01
Supplier:  Abnova
Recombinant protein, tspan8( np_004607, 110 a.a. - 205 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007103-Q01
Supplier:  Abnova