Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, taf4( np_003176, 715 a.a. - 800 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006874-Q01
Supplier:  Abnova
Recombinant protein, taf4( np_003176, 715 a.a. - 800 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006874-Q01
Supplier:  Abnova