Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, abcc8( np_000343, 611 a.a. - 710 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalogus # ABNOH00006833-Q01
Supplier:  Abnova
undefined
Recombinant protein, abcc8( np_000343, 611 a.a. - 710 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalogus # ABNOH00006833-Q01
Supplier:  Abnova