Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, arvcf( np_001661, 863 a.a. - 962 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000421-Q01
Supplier:  Abnova
undefined
Recombinant protein, arvcf( np_001661, 863 a.a. - 962 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000421-Q01
Supplier:  Abnova