Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, clcn3( np_001820.2, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001182-Q01
Supplier:  Abnova
Recombinant protein, clcn3( np_001820.2, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001182-Q01
Supplier:  Abnova