Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, ccne1( np_001229, 311 a.a. - 410 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalogus # ABNOH00000898-Q01
Supplier:  Abnova
Recombinant protein, ccne1( np_001229, 311 a.a. - 410 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalogus # ABNOH00000898-Q01
Supplier:  Abnova