Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, bmp3( np_001192, 373 a.a. - 472 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000651-Q01
Supplier:  Abnova
Recombinant protein, bmp3( np_001192, 373 a.a. - 472 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000651-Q01
Supplier:  Abnova