Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, idh2( np_002159, 354 a.a. - 451 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003418-Q01
Supplier:  Abnova
Recombinant protein, idh2( np_002159, 354 a.a. - 451 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003418-Q01
Supplier:  Abnova