Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, vps52( np_072047.4, 1 a.a. - 723 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006293-P01
Supplier:  Abnova
Recombinant protein, vps52( np_072047.4, 1 a.a. - 723 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006293-P01
Supplier:  Abnova