Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, trim16( np_006461.3, 1 a.a. - 564 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010626-P01
Supplier:  Abnova
Recombinant protein, trim16( np_006461.3, 1 a.a. - 564 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010626-P01
Supplier:  Abnova