Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, mrpl28( np_006419.2, 1 a.a. - 256 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010573-P01
Supplier:  Abnova
Recombinant protein, mrpl28( np_006419.2, 1 a.a. - 256 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010573-P01
Supplier:  Abnova