Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, vdac3( aah56870, 1 a.a. - 284 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007419-P01
Supplier:  Abnova
Recombinant protein, vdac3( aah56870, 1 a.a. - 284 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007419-P01
Supplier:  Abnova