Recombinant protein, prmt2( np_001526.2, 1 a.a. - 433 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003275-P01
Leverancier: Abnova
Recombinant protein, prmt2( np_001526.2, 1 a.a. - 433 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003275-P01
Leverancier: Abnova
Frequently Bought Together
VWR® Facial Tissues
Leverancier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
VWR®, Nitrile Gloves
Leverancier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Leverancier: Merck
Acetone ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Leverancier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Leverancier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Leverancier: VWR Collection
VWR® Serological Pipette, Standard Line
Leverancier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Leverancier: KIMBERLY CLARK