Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, atp5e( aah01690, 1 a.a. - 52 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000514-P01
Supplier:  Abnova
Recombinant protein, atp5e( aah01690, 1 a.a. - 52 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000514-P01
Supplier:  Abnova