Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, trim23( aah22510, 1 a.a. - 575 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000373-P01
Supplier:  Abnova
Recombinant protein, trim23( aah22510, 1 a.a. - 575 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000373-P01
Supplier:  Abnova