Order Entry
Netherlands
ContactUsLinkComponent
Recombinant protein, ckm( aah07462, 1 a.a. - 382 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001158-P01
Supplier:  Abnova
Recombinant protein, ckm( aah07462, 1 a.a. - 382 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001158-P01
Supplier:  Abnova