Order Entry
Netherlands
ContactUsLinkComponent
Systec DX-200: Horizontal, front-loading autoclave Chamber volume (Liter) Total / Nominal 211/200. Chamber dimensions (mm) Ø x dept h: 500 x 1000 1 * 1 items
Catalog # SYST2201
Supplier:  Systec GmbH
Systec DX-200: Horizontal, front-loading autoclave Chamber volume (Liter) Total / Nominal 211/200. Chamber dimensions (mm) Ø x dept h: 500 x 1000 1 * 1 items
Catalog # SYST2201
Supplier:  Systec GmbH
Frequently Bought Together