Ep-CAM (L54) mouse monoclonal IgG2a 100 µg/ml raised against the extracellular domain of Ep-CAM of human origin, with epitope map ping to amino acids 136-265 1 * 1 Each
Catalog # SANTSC-73941
Supplier: Santa Cruz Biotechnology
Frequently Bought Together