Order Entry
Netherlands
ContactUsLinkComponent
InPro 4800/225/Pt1000 pH-ELECTRODE with ring diaphragm made from PTFE Measuring range: 0...14 pH Temperature range: -5...130 °C (operating state) Pressure resistance: 0...13 bar over-pressure 13 bar (130°C) Temperature sensor: Pt1000 Membrane glass: 1 * 1 items
  METN52002127
InPro 4800/225/Pt1000 pH-ELECTRODE with ring diaphragm made from PTFE Measuring range: 0...14 pH Temperature range: -5...130 °C (operating state) Pressure resistance: 0...13 bar over-pressure 13 bar (130°C) Temperature sensor: Pt1000 Membrane glass: 1 * 1 items
  METN52002127
Frequently Bought Together