Order Entry
Netherlands
ContactUsLinkComponent
STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST18 STAINLESS STEEL TANK, 18L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT +5 TO 200C. 1 * 1 items
Catalog # 461-0717
Supplier:  GRANT INSTRUMENTS
STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST18 STAINLESS STEEL TANK, 18L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT +5 TO 200C. 1 * 1 items
Catalog # 461-0717
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together