Iron, powder
Glass bottle for solids, 5.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7439-89-6
- Formula:Fe
- MDL Number:MFCD00010999
- Molecular Weight:55.85 g/mol
- Boiling Pt:3000 °C (1013 hPa)
- Melting Pt:1536 °C
- Density:7.86 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-096-4
- UN:3089
- ADR:4.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



