Order Entry
Netherlands
ContactUsLinkComponent
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST38 STAINLESS STEEL TANK, 38L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST38EU
Supplier:  GRANT INSTRUMENTS
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST38 STAINLESS STEEL TANK, 38L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST38EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together