Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Calibration weights, class F1

5 kg, 25,0 ± mg, Cylindrical test weight without certificate
Catalogus # SARBYCW6538-00
Leverancier:   Sartorius Balances
Calibration weights, class F1
Catalogus # SARBYCW6538-00
Leverancier:  Sartorius Balances
Calibration weights, class F1
Calibration weights, class F1

5 kg, 25,0 ± mg, Cylindrical test weight without certificate

Price
661,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Cylindrical weights which are made up of special stainless steel with a density of 7,950 kg/m³.

 : Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.

 : Cylindrical weights are supplied in high-quality plastic cases that are suitable for use in cleanrooms.

Specificaties
  • Weight:
    5 kg
  • Seq:
    12
  • Tolerance:
    25,0 ± mg
  • Type:
    Cylindrical test weight without certificate