Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Stopper, HDPE, KIMBLE®
Stopper, HDPE, KIMBLE®
13 mm
Catalogus # SCERSP28160R-13
Leverancier:   DWK Life Sciences
Stopper, HDPE, KIMBLE®
Catalogus # SCERSP28160R-13
Leverancier:  DWK Life Sciences
Stopper, HDPE, KIMBLE®
13 mm
Price
34,10 €
Log in voor beschikbaarheid
1 * 6 ST
Product Details & Documents
HDPE, linear, ideal for flasks that are grounded to [ST] stopper dimensions.
  • Yellow and blue stopper with a closed bottom
  • Enlarged flange is designed to protect the neck if the glassware is tipped over
Specificaties
  • Ø bottom:
    13 mm
  • Seq:
    02
Aanbevelingen