Order Entry
Netherlands
ContactUsLinkComponent
Semi-automated vacuum controller V-850 with a 1/4 VGA LCD-display to control the vacuum by a valve or by the new speed control techn ology with the Vacuum Pump V-700/V-710. The cont 1 * 1 items
Catalog # BUCH047293
Supplier:  BUCHI
Semi-automated vacuum controller V-850 with a 1/4 VGA LCD-display to control the vacuum by a valve or by the new speed control techn ology with the Vacuum Pump V-700/V-710. The cont 1 * 1 items
Catalog # BUCH047293
Supplier:  BUCHI
Frequently Bought Together