Order Entry
Netherlands
ContactUsLinkComponent
pe canister capacity: 10 lcapacity: approx. 20 l dimensions: 298 x 198 x 499 (w x d x h mm) filler dia: 56 mm colour of material: bl ack, incl. screw cap acc. to trbf 60. administration as package for transport of dangerous goods of packing group ii 1 * 1 items
Catalog # DUEH5.09.020
Supplier:  DUPERTHAL
pe canister capacity: 10 lcapacity: approx. 20 l dimensions: 298 x 198 x 499 (w x d x h mm) filler dia: 56 mm colour of material: bl ack, incl. screw cap acc. to trbf 60. administration as package for transport of dangerous goods of packing group ii 1 * 1 items
Catalog # DUEH5.09.020
Supplier:  DUPERTHAL
Frequently Bought Together