Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FORMALDEHYDE 4% 5 LTR CUBITAINER 1 * 5 L
Catalog # VWRK4078.9005
Supplier:
KLINIPATH
Certificates
Expand image
FORMALDEHYDE 4% 5 LTR CUBITAINER 1 * 5 L
Catalog # VWRK4078.9005
Supplier:
KLINIPATH
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
IFU Buffered Formaldehyde 4% mv.pdf
Product specification
spec_VWRK4078-9005
spec_VWRK4078.9005
Documentation
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
VWR® Overalls, SMS
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR®, Cover Glasses, Round
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
cDNA synthesis kits, qScript™
Supplier: Quantabio