OmniPur Sodium Dodecyl Sulfate 1 * 500 g
Catalog # CALB7910-500GM
Supplier: Merck Millipore (Calbiochem)
OmniPur Sodium Dodecyl Sulfate 1 * 500 g
Catalog # CALB7910-500GM
Supplier: Merck Millipore (Calbiochem)
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Supplier: Merck
Diisopropyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
9-Fluorenylmethyl chloroformate (Fmoc chloride), Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.9% ACS
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
1-Butanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals