Order Entry
Netherlands
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase 10% SP-1000, matrix 80/100 SUPELCOPORT support, L x O.D. x I.D. 20.0 ft (6.1 m ) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12719.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase 10% SP-1000, matrix 80/100 SUPELCOPORT support, L x O.D. x I.D. 20.0 ft (6.1 m ) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12719.
Supplier:  Merck
Frequently Bought Together