SulfoLink™ Coupling Resin
SulfoLink™ coupling resin, Liquid, 250 ml
PIER20404
SulfoLink™ coupling resin is porous, crosslinked, 6% beaded agarose that has been activated with iodoacetyl groups for covalent immobilisation of cysteine-peptides and other sulfhydryl molecules.
- Iodoacetyl groups react specifically with sulhydryls (-SH) to form irreversible thioether bonds
- Spin columns increase protocol speed; prepare and couple samples in 2 hours (peptides) to 3,5 hours (proteins)
- Flexible coupling conditions - use pH 7,5 to 9,0 aqueous buffers, organic solvent (eg. 20% DMSO) or denaturant (guanidine·HCl), as needed for protein or peptide solubility during coupling reaction
- Easy-to-follow protocols for sample preparation, immobilisation, and affinity purification
- High capacity - immobilise 1 to 2 mg of peptide or 2 to 20 mg of protein per 2 ml column
: Supplied as a 50% slurry in storage buffer (10 mM EDTA-Na, 0.05% NaN₃, 50% glycerol).
- Pk:250 ml
- Description:SulfoLink™ coupling resin
- Type:Liquid
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



