Order Entry
Netherlands
ContactUsLinkComponent
25 LITRE DIGITAL ULTRASONIC BATH, TEMP RANGE: AMBIENT+5#C to 70#C. MAINS SUPPLY: 230V AC 50/60Hz. INCLUDES STAINLESS STEEL BASKET A ND ABS PLASTIC LID, 1 BOTTLE OF M2 SOLUTION, DRAIN HOSE, SD CARD, SD CARD READER AND EUCORDSET 1 * 1 items
Catalog # GRANXUB25EURO
Supplier:  GRANT INSTRUMENTS
25 LITRE DIGITAL ULTRASONIC BATH, TEMP RANGE: AMBIENT+5#C to 70#C. MAINS SUPPLY: 230V AC 50/60Hz. INCLUDES STAINLESS STEEL BASKET A ND ABS PLASTIC LID, 1 BOTTLE OF M2 SOLUTION, DRAIN HOSE, SD CARD, SD CARD READER AND EUCORDSET 1 * 1 items
Catalog # GRANXUB25EURO
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together